missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dithiothreitol (White Crystals or Powder/Electrophoresis), Fisher BioReagents™
Dithiothreitol, >-99%, C4H10O2S2, CAS Number-3483-12-3, 1,4-dithio-dl-threitol, 1,4-dithiothreitol, 2s,3s-1,4-dimercaptobutane-2,3-diol, d-1,4-dithiothreitol, d-dtt, dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, dtt, threo-1,4-dimercapto-2,3-butanediol, 5g, 0.04 AU max., >-99 %
47.80 €
Chemical Identifiers
CAS | 3483-12-3 |
---|---|
Molecular Formula | C4H10O2S2 |
Molecular Weight (g/mol) | 154.24 |
InChI Key | VHJLVAABSRFDPM-IMJSIDKUSA-N |
Synonym | dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt |
PubChem CID | 446094 |
ChEBI | CHEBI:42170 |
IUPAC Name | (2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol |
SMILES | C(C(C(CS)O)O)S |
Description
Dithiothreitol, >-99%, C4H10O2S2, CAS Number-3483-12-3, 1,4-dithio-dl-threitol, 1,4-dithiothreitol, 2s,3s-1,4-dimercaptobutane-2,3-diol, d-1,4-dithiothreitol, d-dtt, dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, dtt, threo-1,4-dimercapto-2,3-butanediol, 5g, 0.04 AU max., >-99 %
Dithiothreitol is used to reduce disulfide bonds in proteins and peptides and is added in low concentrations as an enzyme stabilizer during protein purification and enzymatic reactions.Chemical Identifiers
3483-12-3 | |
154.24 | |
dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt | |
CHEBI:42170 | |
C(C(C(CS)O)O)S |
C4H10O2S2 | |
VHJLVAABSRFDPM-IMJSIDKUSA-N | |
446094 | |
(2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol |
Specifications
3483-12-3 | |
38°C | |
5.1 | |
≥99 % | |
HSCH2CH(OH)CH(OH)CH2SH | |
dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt | |
C(C(C(CS)O)O)S | |
154.24 | |
CHEBI:42170 | |
≥99% | |
Powder or Solid |
0.04 AU max. | |
White | |
125°C | |
C4H10O2S2 | |
15, 3419 | |
VHJLVAABSRFDPM-IMJSIDKUSA-N | |
(2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol | |
446094 | |
154.24 | |
DNase-, RNase- and Protease-Free | |
Dithiothreitol |
Safety and Handling
Recommended Storage : -20°C
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- DL-1,4-Dithiothreitol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Dithiothreitol
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Dithiothreitol (White Crystals or Powder/Electrophoresis), Fisher BioReagents™