missing translation for 'onlineSavingsMsg'
Learn More

Dithranol, 99%, MP Biomedicals™

Dithranol ≥99%, Yellow Powder

132.00 €

Chemical Identifiers

CAS 1143-38-0
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.23
MDL Number MFCD00053409
InChI Key NUZWLKWWNNJHPT-UHFFFAOYSA-N
PubChem CID 2202
ChEBI CHEBI:37510
IUPAC Name 1,8-dihydroxy-9,10-dihydroanthracen-9-one
SMILES OC1=CC=CC2=C1C(=O)C1=C(O)C=CC=C1C2
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11424742
View Documents Promotion Details
MP Biomedicals
0219150291
1 g
132.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Dithranol, a potential anti-psoriasis medication, may be used to study its safety, efficacy, metabolism and pharmacological profile.
  • It may be used to study is physicochemical properties, as a reference compound and to develop effective drug delivery vehicles.
  • It may be used as a matrix for matrix assisted laser desorption/ionization imaging on a Fourier transform ion cyclotron resonance mass spectrometer.
  • Anthralin is a leukotriene biosynthesis inhibitor.
  • Inhibits LTB4 ω-oxidation and disrupts mitochondria function.
  • Anthralin is used in the treatment of psoriasis, as a fungicide, in the treatment of ringworm infections and in chronic dermatoses.
Specifications

Chemical Identifiers

1143-38-0
226.23
NUZWLKWWNNJHPT-UHFFFAOYSA-N
CHEBI:37510
OC1=CC=CC2=C1C(=O)C1=C(O)C=CC=C1C2
C14H10O3
MFCD00053409
2202
1,8-dihydroxy-9,10-dihydroanthracen-9-one

Specifications

1143-38-0
≥99%
MFCD00053409
NUZWLKWWNNJHPT-UHFFFAOYSA-N
1,8-dihydroxy-9,10-dihydroanthracen-9-one
2202
226.23
Dithranol
Orange-Yellow or Yellow
C14H10O3
anthralin, dithranol, 1,8-dihydroxyanthrone, 1,8-dihydroxy-9-anthrone, chrysodermol, cigthranol, batridol, cignolin, psoriacid-stift, 9 10h-anthracenone, 1,8-dihydroxy
OC1=CC=CC2=C1C(=O)C1=C(O)C=CC=C1C2
226.23
CHEBI:37510
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 214-538-0

Recommended Storage : Store at Room Temperature (15°to 30°C)

Product Identifier
  • dithranol

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : dithranol

SDS
Documents

Documents

Product Certifications