missing translation for 'onlineSavingsMsg'
Learn More

Divinyltin dichloride, Thermo Scientific™

Product Code. 11374547
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7532-85-6
Molecular Formula C4H8Cl2Sn
Molecular Weight (g/mol) 245.72
MDL Number MFCD00058728
InChI Key OFJXSWMLVIFLKC-UHFFFAOYSA-N
Synonym divinyltin dichloride, dichlorodivinylstannane, stannane, dichlorodiethenyl, dichloro divinyl stannane, divinyltindichloride, acmc-20al40, dichloro-bis ethenyl stannane
PubChem CID 82033
IUPAC Name dichloro-bis(ethenyl)stannane
SMILES Cl.Cl.C=C[SnH2]C=C

Specifications

Boiling Point 54°C to 56°C (3 mmHg)
Quantity 2 g
UN Number UN2788
Sensitivity Moisture sensitive
Formula Weight 243.69
Physical Form Liquid
Chemical Name or Material Divinyltin dichloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'