missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-1,4-Dithiothreitol, 99+%, for molecular biology, DNAse, RNAse and Protease free

Product Code. 10551754
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass Bottle
Plastic Bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3483-12-3
ChEBI CHEBI:42170

Specifications

Absorbance (0.1 M in water), (0.1 M in water) at 0nm, 0.1 max. at 280nm, 0.5 max. at 260nm
Melting Point 39.0°C to 44.0°C
Color Colorless to Yellow
Boiling Point 125.0°C to 130.0°C (2.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass Bottle
Molecular Formula C4H10O2S2
Linear Formula HSCH2CH(OH)CH(OH)CH2SH
MDL Number MFCD00004877
Quantity 1 g
Beilstein 01, III, 2360
Merck Index 15, 3419
Synonym dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt
InChI Key VHJLVAABSRFDPM-IMJSIDKUSA-N
SMILES C(C(C(CS)O)O)S
IUPAC Name (2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol
Molecular Weight (g/mol) 154.24
PubChem CID 446094
Formula Weight 154.24
Percent Purity ≥99%
Grade Molecular Biology
Physical Form Powder
Chemical Name or Material DL-1, 4-Dithiothreitol, DNase, RNase, Protease free
Show More Show Less
compliance-icons
Product Identifier
  • DL-1,4-Dithiothreitol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.