missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-1,4-Dithiothreitol, For Biochemistry, 1M solution in water

31.86 € - 397.98 €

Specifications

Chemical Name or Material DL-1, 4-Dithiothreitol
Color Colorless
Name Note 1M Solution in water, For molecular biology
CAS 3483-12-3, 7732-18-5
Concentration 0.90 to 1.10M
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10162994
View Documents
Thermo Scientific Chemicals
426380100
10 mL Glass bottle
31.86 €
10mL
Please to purchase this item. Need a web account? Register with us today!
10674545
View Documents
Thermo Scientific Chemicals
426380500
50 mL Glass bottle
89.55 €
50mL
Please to purchase this item. Need a web account? Register with us today!
10215054
View Documents
Thermo Scientific Chemicals
426382500
250 mL Glass bottle
397.98 €
250mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For biochemistry
Specifications

Specifications

DL-1, 4-Dithiothreitol
1M Solution in water, For molecular biology
0.90 to 1.10M
15, 3419
Biochemical
1.04
Colorless
3483-12-3, 7732-18-5
1.0400g/mL
154.24
Glass bottle
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Product Identifier
  • DL-Dithiothreitol, 1M solution in water

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only