missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Octanol, 97%

Product Code. 10155253 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
100 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
100mL
2.5 L
5mL
This item is not returnable. View return policy

Product Code. 10155253

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123-96-6 | C8H18O | 130.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-96-6
MDL Number MFCD00004591
InChI Key SJWFXCIHNDVPSH-UHFFFAOYSA-N
Synonym 2-octanol, 2-octyl alcohol, hexylmethylcarbinol, 2-hydroxyoctane, sec-caprylic alcohol, 1-methylheptanol, methylhexylcarbinol, 1-methylheptyl alcohol, dl-2-octanol, n-octan-2-ol
PubChem CID 20083
ChEBI CHEBI:37869
IUPAC Name octan-2-ol
SMILES CCCCCCC(C)O

Specifications

Melting Point -38.0°C
Color Colorless to Yellow
Density 0.8100g/mL
Boiling Point 174.0°C to 181.0°C
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C8H18O
Linear Formula CH3(CH2)5CH(OH)CH3
Refractive Index 1.4250 to 1.4270
Quantity 2.5 L
Beilstein 01, 419
Fieser 01,109
Merck Index 15, 6841
Specific Gravity 0.81
Solubility Information Solubility in water: 1g/L (20°C). Other solubilities: miscible with alcohol and ether, soluble in most organic solvents
Molecular Weight (g/mol) 130.23
Formula Weight 130.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material DL-2-Octanol
Show More Show Less
compliance-icons
Product Identifier
  • DL-2-Octanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.