missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-6-Hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate, 96%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Specifications
Specifications
CAS Min % | 95.0 |
CAS Max % | 100.0 |
Melting Point | 256°C to 260°C |
Color | White to Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 95% min. (HPLC) |
Packaging | Glass bottle |
Quantity | 250 mg |
IUPAC Name | 6-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid;hydrate |
Formula Weight | 211.22 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 7
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DL-6-Hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate, 96%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction