missing translation for 'onlineSavingsMsg'
Learn More
DL-Alanine 99%, Thermo Scientific Chemicals Product Code.: 10183490

DL-Alanine 99%, Thermo Scientific Chemicals

Product Code. 10183490
500 g, Plastic Bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
This item is not returnable. View return policy

Product Code. 10183490

Brand: Thermo Scientific Acros 159095000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 302-72-7
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00064408
InChI Key QNAYBMKLOCPYGJ-UHFFFAOYNA-N
Synonym dl-alanine, alanine, alanine, dl, dl-alpha-alanine, d,l-alanine, dl-2-aminopropionic acid, r,s-alanine, dl-2-aminopropanoic acid, h-dl-ala-oh, +--alanine
PubChem CID 602
ChEBI CHEBI:16449
IUPAC Name 2-aminopropanoic acid
SMILES CC(N)C(O)=O

Specifications

Melting Point 272.0°C to 275.0°C
Ammonium 200ppm max.
Arsenic (As) 1ppm max.
pH 5.5 to 7 (2.5% soln. at 25°C)
Color White
Chlorine (Cl) 200ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Assay Percent Range 99%
Loss on Drying 0.2% max. (105°C, 3 hrs)
Other Amino Acids 0.5% max. (TLC)
Packaging Plastic Bottle
Linear Formula CH3CH(NH2)CO2H
Beilstein 04, 387
Sulfate 300ppm max.
Merck Index 15, 197
Transmittance 95% min. (c=10, H2O, 430nm) 1cm cell
Sulfated Ash 0.1% max.
Solubility Information Solubility in water: 156g/L in water (20°C). Other solubilities: slightly soluble in alcohol, insoluble in ether and organic solvents, soluble in mineral acids, soluble in ethanol 0.0087g/100g (25°C)
Sublimation Point >190 °C
Quantity 500 g
Formula Weight 89.09
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material DL-Alanine, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466339 Certificate TypeCertificate of Analysis Date02/10/2024 Catalog Number159091000,159095000
Lot NumberA0458868 Certificate TypeCertificate of Analysis Date05/01/2024 Catalog Number159091000,159095000
Lot NumberA0394758 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number159091000,159095000,159090025
Lot NumberA0411341 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number159091000,159095000,159090025
Lot NumberA0435212 Certificate TypeCertificate of Analysis Date20/10/2021 Catalog Number159091000,159095000,159090025

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DL-Alanine 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.