missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals DL-Asparagine monohydrate, 98%
Click to view available options
Quantity:
100 g
500 g
Description
DL-Asparagine monohydrate is used as a pharmaceutical intermediate. It is used to prepare DL-asparaginium nitrate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
DL-Asparagine monohydrate is used as a pharmaceutical intermediate. It is used to prepare DL-asparaginium nitrate.
Solubility
Soluble in water, acids, and alkalis.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
DL-Asparagine monohydrate is used as a pharmaceutical intermediate. It is used to prepare DL-asparaginium nitrate.
Solubility
Soluble in water, acids, and alkalis.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Specifications
Specifications
Melting Point | ∼220°C (decomposition) |
Beilstein | 6234992 |
Solubility Information | Soluble in water,acids,and alkalis. |
IUPAC Name | 2,4-diamino-4-oxobutanoic acid;hydrate |
Quantity | 500 g |
Formula Weight | 150.14 (132.12 Anhydrous) |
Percent Purity | 98% |
Chemical Name or Material | DL-Asparagine monohydrate |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific Chemicals DL-Asparagine monohydrate, 98% >
Spot an opportunity for improvement?Share a Content Correction