Learn More
DL-Buthionine (S,R)-sulfoximine, 99%, Thermo Scientific Chemicals
137.00 € - 1007.00 €
Chemical Identifiers
CAS | 5072-26-4 |
---|---|
Molecular Formula | C8H18N2O3S |
Molecular Weight (g/mol) | 222.30 |
MDL Number | MFCD00070309 |
InChI Key | KJQFBVYMGADDTQ-UHFFFAOYNA-N |
Synonym | buthionine sulfoximine, dl-buthionine-s,r-sulfoximine, buthionine sulfoxamine, buthione sulfoximine, buthionine-s,r-sulfoximine, 2-amino-4-s-butylsulfonimidoyl butanoic acid, butionine sulfoximine, buthionine sulphoximine, 2-amino-4-butylsulfonimidoyl butanoic acid, butanoic acid, 2-amino-4-s-butylsulfonimidoyl |
PubChem CID | 21157 |
ChEBI | CHEBI:28714 |
IUPAC Name | 2-amino-4-(butylsulfonimidoyl)butanoic acid |
SMILES | CCCCS(=N)(=O)CCC(N)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10480151
|
Thermo Scientific Acros
309475000 |
500 mg | Glass bottle |
137.00 €
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10646554
|
Thermo Scientific Acros
309470050 |
5 g | Glass bottle |
1007.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5072-26-4 | |
222.30 | |
KJQFBVYMGADDTQ-UHFFFAOYNA-N | |
21157 | |
2-amino-4-(butylsulfonimidoyl)butanoic acid |
C8H18N2O3S | |
MFCD00070309 | |
buthionine sulfoximine, dl-buthionine-s,r-sulfoximine, buthionine sulfoxamine, buthione sulfoximine, buthionine-s,r-sulfoximine, 2-amino-4-s-butylsulfonimidoyl butanoic acid, butionine sulfoximine, buthionine sulphoximine, 2-amino-4-butylsulfonimidoyl butanoic acid, butanoic acid, 2-amino-4-s-butylsulfonimidoyl | |
CHEBI:28714 | |
CCCCS(=N)(=O)CCC(N)C(O)=O |
Specifications
5072-26-4 | |
White | |
99% | |
CH3(CH2)3S(O)(=NH)CH2CH2CH(NH2)CO2H | |
15, 1528 | |
Solubility in water: soluble | |
CCCCS(=N)(=O)CCC(N)C(O)=O | |
222.30 | |
CHEBI:28714 | |
99% | |
DL-Buthionine (S, R)-sulfoximine, 99% |
215°C | |
Authentic | |
C8H18N2O3S | |
MFCD00070309 | |
buthionine sulfoximine, dl-buthionine-s,r-sulfoximine, buthionine sulfoxamine, buthione sulfoximine, buthionine-s,r-sulfoximine, 2-amino-4-s-butylsulfonimidoyl butanoic acid, butionine sulfoximine, buthionine sulphoximine, 2-amino-4-butylsulfonimidoyl butanoic acid, butanoic acid, 2-amino-4-s-butylsulfonimidoyl | |
KJQFBVYMGADDTQ-UHFFFAOYNA-N | |
2-amino-4-(butylsulfonimidoyl)butanoic acid | |
21157 | |
222.31 | |
Fluffy Powder |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'rtecsNumber' : EK7713435
RUO – Research Use Only