missing translation for 'onlineSavingsMsg'
Learn More

DL-Glutamic acid, 95%, Thermo Scientific Chemicals

Product Code. 15439558
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Its salt form (glutamate) is an important neurotransmitter that plays a key role in long-term potentiation and is important for learning and memory. Glutamic Acid is also a key molecule in cellular metabolism. DL-Glutamic acid is used ito study its mechanisms of crystal formation and self-assembly on surfaces and copolymer development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Its salt form (glutamate) is an important neurotransmitter that plays a key role in long-term potentiation and is important for learning and memory. Glutamic Acid is also a key molecule in cellular metabolism. DL-Glutamic acid is used ito study its mechanisms of crystal formation and self-assembly on surfaces and copolymer development.

Solubility
Soluble in water (miscible), HCl, ether (sparingly), ethanol (sparingly), and petroleum ether (sparingly).

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-65-2
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
MDL Number MFCD00063113
InChI Key WHUUTDBJXJRKMK-UHFFFAOYNA-N
Synonym dl-glutamic acid, glutamic acid, glutamic acid, dl, glutaminsaeure, glutamic acid dl-form, dl-glutamicacid, dl-glu, +--glutamic acid, 2-aminoglutaric acid, glutamate
PubChem CID 611
ChEBI CHEBI:18237
IUPAC Name 2-aminopentanedioic acid
SMILES NC(CCC(O)=O)C(O)=O

Specifications

Beilstein 1723799
Merck Index 14,4469
Solubility Information Soluble in water (miscible),HCl,ether (sparingly),ethanol (sparingly),and petroleum ether (sparingly).
Quantity 25 g
Formula Weight 147.13
Percent Purity 95%
Chemical Name or Material DL-Glutamic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'