Learn More
Thermo Scientific Chemicals DL-Histidine monohydrochloride monohydrate, 99%
Description
DL-Histidine monohydrochloride monohydrate is the mixed isomer of D- and L-histidine. It may be used to evaluate the efficiency of amino acid chiral ligand exchange media and systems and in running buffers for capillary electrophoresis. It is also an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
DL-Histidine monohydrochloride monohydrate is the mixed isomer of D- and L-histidine. It may be used to evaluate the efficiency of amino acid chiral ligand exchange media and systems and in running buffers for capillary electrophoresis. It is also an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Specifications
Specifications
Melting Point | ∼250°C (decomposition) |
Solubility Information | Insoluble in water. |
Quantity | 25 g |
Formula Weight | 209.63 (191.62 Anhydrous) |
Percent Purity | 99% |
Chemical Name or Material | DL-Histidine monohydrochloride monohydrate |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.