missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Homoserine, 99%

Product Code. 10636252
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1927-25-9
Color White
Assay Percent Range 99%
Molecular Formula C4H9NO3
Infrared Spectrum Authentic
Linear Formula HOCH2CH2CH(NH2)CO2H
MDL Number MFCD00002618,MFCD00077786
Beilstein 04, 514
Merck Index 14, 4739
Packaging Glass Bottle
Synonym dl-homoserine, homoserine, h-dl-hoser-oh, d,l-homoserine, h-d-homoser-oh, butyric acid, 2-amino-4-hydroxy-, dl, 2-amino-4-hydroxy-butanoic acid, dl-2-amino-4-hydroxybutyric acid, d-homoserin, h-dl-hse-oh
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in ethanol
InChI Key UKAUYVFTDYCKQA-VKHMYHEASA-N
SMILES N[C@@H](CCO)C(O)=O
IUPAC Name 2-amino-4-hydroxybutanoic acid
Molecular Weight (g/mol) 119.12
PubChem CID 779
ChEBI CHEBI:30653
Formula Weight 119.12
Percent Purity 98.5 to 101%
Quantity 5 g
Physical Form Fine Crystalline Powder or Needles
Chemical Name or Material DL-Homoserine, 99%
Show More Show Less
Product Identifier
  • DL-Homoserine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'