missing translation for 'onlineSavingsMsg'
Learn More

DL-Isocitric acid, trisodium salt hydrate, 95%

Product Code. 10388050 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 10388050

Brand: Thermo Scientific undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1637-73-6 | C6H5Na3O7 | 258.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1637-73-6
Molecular Formula C6H5Na3O7
Molecular Weight (g/mol) 258.07
MDL Number MFCD00149284
InChI Key HWMVXEKEEAIYGB-UHFFFAOYNA-K
Synonym dl-isocitric acid, isocitric acid trisodium salt, sodium isocitrate, dl-isocitric acid trisodium salt hydrate, dl-isocitric acid trisodium salt, trisodium 1-hydroxypropane-1,2,3-tricarboxylate, trisodium isocitrate, tripotassium isocitrate, isocitric acid, dl, dl-isocitric acid, trisodium salt hydrate
PubChem CID 168942
IUPAC Name trisodium;1-hydroxypropane-1,2,3-tricarboxylate
SMILES [Na+].[Na+].[Na+].OC(C(CC([O-])=O)C([O-])=O)C([O-])=O

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula NaO2CCH2CH(CO2Na)CH(OH)CO2Na·xH2O
Quantity 1 g
Beilstein 03, 555
Solubility Information Solubility in water: soluble in water
Formula Weight 258.07
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material DL-Isocitric acid, trisodium salt hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.