missing translation for 'onlineSavingsMsg'
Learn More

DL-Lactic acid, 80-85% aq. soln., Thermo Scientific Chemicals

Product Code. 15419009
Click to view available options
Quantity:
250 g
1000 g
2500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

DL-Lactic acid is used to produce serial products or widely used in food, vintage, beverage, drugs, polymerization, textile, leather, tobacco, feed, plastic chemicals, pesticide, polymer solution and other industry. Lactic acid is also used as an acidifying agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water and in ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-21-5
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00004520
InChI Key JVTAAEKCZFNVCJ-UHFFFAOYNA-N
Synonym lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum
PubChem CID 612
ChEBI CHEBI:78320
IUPAC Name 2-hydroxypropanoic acid
SMILES CC(O)C(O)=O

Specifications

Boiling Point 122°C (15 mmHg)
Beilstein 1209341
Merck Index 145336
Solubility Information Soluble in water and in ethanol.
Formula Weight 90.08
Concentration or Composition (by Analyte or Components) 80 to 85% aq. Soln.
Density 1.209
Quantity 250 g
Flash Point >110°C (230°F)
Refractive Index 1.423
Chemical Name or Material DL-Lactic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'