Learn More
DL-Lactic acid, 80-85% aq. soln.
Description
DL-Lactic acid is used to produce serial products or widely used in food, vintage, beverage, drugs, polymerization, textile, leather, tobacco, feed, plastic chemicals, pesticide, polymer solution and other industry. Lactic acid is also used as an acidifying agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Soluble in water and in ethanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Specifications
| Boiling Point | 122°C (15 mmHg) |
| Beilstein | 1209341 |
| Merck Index | 145336 |
| Solubility Information | Soluble in water and in ethanol. |
| Formula Weight | 90.08 |
| Concentration or Composition (by Analyte or Components) | 80 to 85% aq. Soln. |
| Density | 1.209 |
| Quantity | 1000 g |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.423 |
| Show More |
Safety and Handling
- DL-Lactic acid, 80-85% aqueous solution
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 C
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- EUH071-Corrosive to the respiratory tract.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Lactic acid
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.