Learn More
DL-Mandelic acid, 99+%, Thermo Scientific Chemicals
33.90 € - 654.00 €
Chemical Identifiers
CAS | 90-64-2 |
---|---|
Molecular Formula | C8H8O3 |
Molecular Weight (g/mol) | 152.149 |
InChI Key | IWYDHOAUDWTVEP-UHFFFAOYSA-N |
Synonym | mandelic acid, dl-mandelic acid, phenylglycolic acid, amygdalic acid, almond acid, uromaline, p-mandelic acid, racemic mandelic acid, paramandelic acid, rs-mandelic acid |
PubChem CID | 1292 |
ChEBI | CHEBI:35825 |
IUPAC Name | 2-hydroxy-2-phenylacetic acid |
SMILES | C1=CC=C(C=C1)C(C(=O)O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10625942
|
Thermo Scientific Acros
125311000 |
100 g | Plastic bottle |
33.90 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10745771
|
Thermo Scientific Acros
125312500 |
250 g | Plastic bottle |
56.10 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10416750
|
Thermo Scientific Acros
125310010 |
1 kg | Plastic bottle |
138.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11975071
|
Thermo Scientific Acros
125310050 |
5 kg | Plastic Drum |
654.00 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
90-64-2 | |
152.149 | |
mandelic acid, dl-mandelic acid, phenylglycolic acid, amygdalic acid, almond acid, uromaline, p-mandelic acid, racemic mandelic acid, paramandelic acid, rs-mandelic acid | |
CHEBI:35825 | |
C1=CC=C(C=C1)C(C(=O)O)O |
C8H8O3 | |
IWYDHOAUDWTVEP-UHFFFAOYSA-N | |
1292 | |
2-hydroxy-2-phenylacetic acid |
Specifications
90-64-2 | |
White | |
C8H8O3 | |
mandelic acid, dl-mandelic acid, phenylglycolic acid, amygdalic acid, almond acid, uromaline, p-mandelic acid, racemic mandelic acid, paramandelic acid, rs-mandelic acid | |
C1=CC=C(C=C1)C(C(=O)O)O | |
152.149 | |
CHEBI:35825 | |
≥99.0% (Acid) | |
Crystals or Crystalline Powder |
117°C to 121°C | |
99+% | |
C6H5CH(OH)CO2H | |
IWYDHOAUDWTVEP-UHFFFAOYSA-N | |
2-hydroxy-2-phenylacetic acid | |
1292 | |
152.15 | |
Authentic | |
DL-Mandelic acid, 99+% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
missing translation for 'einecsNumber' : 202-007-6
missing translation for 'rtecsNumber' : OO6300000
missing translation for 'tsca' : TSCA
- DL-Mandelic acid
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only