missing translation for 'onlineSavingsMsg'
Learn More

DL-Mercaptosuccinic acid, 99%

Product Code. 10574051 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10574051

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 70-49-5 | C4H6O4S | 150.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70-49-5
Molecular Formula C4H6O4S
Molecular Weight (g/mol) 150.15
InChI Key NJRXVEJTAYWCQJ-UHFFFAOYSA-N
Synonym mercaptosuccinic acid, thiomalic acid, 2-thiomalic acid, 2-mercaptosuccinic acid, monomercaptosuccinic acid, butanedioic acid, mercapto, malic acid, 2-thio, mercaptosuccinate, 2-sulfanylsuccinic acid, dl-mercaptosuccinic acid
PubChem CID 6268
ChEBI CHEBI:38705
IUPAC Name 2-sulfanylbutanedioic acid
SMILES C(C(C(=O)O)S)C(=O)O

Specifications

Melting Point 152°C to 155°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula HO2CCH2CH(SH)CO2H
Quantity 100 g
Beilstein 03,439
Merck Index 15,9496
Solubility Information Solubility in water: 150g/L (20°C). Other solubilities: 500g/L ethanol (20°C),soluble in acetone,moderately soluble in ether,chloroform,practically insoluble in benzene
Formula Weight 150.15
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material DL-Mercaptosuccinic acid
Show More Show Less
compliance-icons
Product Identifier
  • DL-Mercaptosuccinic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : DL-Mercaptosuccinicacid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.