missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals DL-Methionine sulfoxide, 98+% Product Code.: 11459226

Thermo Scientific Chemicals DL-Methionine sulfoxide, 98+%

Product Code. 11459226
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11459226

Brand: Thermo Scientific Chemicals A18081.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

DL-Methionine sulfoxide is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Methionine sulfoxide is used as pharmaceutical intermediates.

Solubility
Soluble in water and Hcl.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62697-73-8
Molecular Formula C5H11NO3S
Molecular Weight (g/mol) 165.21
MDL Number MFCD00002620
InChI Key QEFRNWWLZKMPFJ-UHFFFAOYNA-N
Synonym dl-methionine sulfoxide, methionine sulfoxide, 2-amino-4-methylsulfinyl butanoic acid, dl-methioninesulfoxide, methionine, s-oxide, uethionine, s-oxide, butanoic acid, 2-amino-4-methylsulfinyl, 2-amino-4-methylsulfinyl butyric acid, 2-amino-4-methanesulfinylbutanoic acid, butyric acid, 2-amino-4-methylsulfinylShow MoreShow Less
PubChem CID 847
ChEBI CHEBI:49033
SMILES CS(=O)CCC(N)C(O)=O

Specifications

Melting Point ∼230°C (decomposition)
Solubility Information Soluble in water and Hcl.
IUPAC Name 2-amino-4-methanesulfinylbutanoic acid
Quantity 25 g
Formula Weight 165.21
Percent Purity ≥98%
Chemical Name or Material DL-Methionine sulfoxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10248158 Certificate TypeCertificate of Analysis Date17/10/2023 Catalog NumberA18081.06,A18081.14
Lot Number10247021 Certificate TypeCertificate of Analysis Date20/06/2023 Catalog NumberA18081,A18081.06,A18081.14
Lot Number10227644 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18081.06,A18081.14
Lot NumberC4460A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18081.06,A18081.14
Lot Number10125671 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18081.06,A18081.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals DL-Methionine sulfoxide, 98+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.