missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Phenylglycinol, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
A chiral arylalkylamine used as organocatalysts. Used in the synthesis of chiral diamines also in determination of chiral recognition properties of novel fluorescent chemosensors for amino acid. an efficient and practical synthesis of l-. alpha.-amino acids using (r)-phenylglycinol as a chiral auxiliary.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A chiral arylalkylamine used as organocatalysts. Used in the synthesis of chiral diamines also in determination of chiral recognition properties of novel fluorescent chemosensors for amino acid. an efficient and practical synthesis of l-. alpha.-amino acids using (r)-phenylglycinol as a chiral auxiliary.
Solubility
Soluble in chloroform, methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
A chiral arylalkylamine used as organocatalysts. Used in the synthesis of chiral diamines also in determination of chiral recognition properties of novel fluorescent chemosensors for amino acid. an efficient and practical synthesis of l-. alpha.-amino acids using (r)-phenylglycinol as a chiral auxiliary.
Solubility
Soluble in chloroform, methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
Melting Point | 49°C to 53°C |
Color | White |
Beilstein | 971219 |
Solubility Information | Soluble in chloroform,methanol. |
Quantity | 25 g |
Formula Weight | 137.18 |
Percent Purity | 95% |
Physical Form | Solid |
Chemical Name or Material | DL-Phenylglycinol |
Safety and Handling
Product Identifier
- DL-Phenylglycinol
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : (.+-.)-2-Amino-2-phenylethan-1-ol
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'