missing translation for 'onlineSavingsMsg'
Learn More
DL-Proline, 99%, Thermo Scientific Chemicals Product Code.: 11484504

DL-Proline, 99%, Thermo Scientific Chemicals

Product Code. 11484504
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11484504

Brand: Thermo Scientific Alfa Aesar A13137.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

DL-Proline is used as an agonist of glycine receptors. It acts as a catalyst for aldol condensation. It is an essential component of collagen. It plays a significant role in the immune system and strengthening of heart muscles. It is an osmoprotectant and therefore is used in many pharmaceutical, biotechnological applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Proline is used as an agonist of glycine receptors. It acts as a catalyst for aldol condensation. It is an essential component of collagen. It plays a significant role in the immune system and strengthening of heart muscles. It is an osmoprotectant and therefore is used in many pharmaceutical, biotechnological applications.

Solubility
Soluble in water and absolute alcohol. Insoluble in ether, butanol and isopropanol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-36-9
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD00005250
InChI Key ONIBWKKTOPOVIA-UHFFFAOYSA-N
Synonym dl-proline, proline, h-dl-pro-oh, proline, dl, 2-pyrrolidine carboxylic acid, hpro, l-proline, homopolymer, +/--pyrrolidine-2-carboxylic acid, prolin, chembl72275Show MoreShow Less
PubChem CID 614
ChEBI CHEBI:26271
IUPAC Name pyrrolidine-2-carboxylic acid
SMILES C1CC(NC1)C(=O)O

Specifications

Melting Point ∼209°C (decomposition)
Beilstein 80809
Merck Index 14,7780
Solubility Information Soluble in water and absolute alcohol. Insoluble in ether,butanol and isopropanol.
Quantity 25 g
Formula Weight 115.13
Percent Purity 99%
Chemical Name or Material DL-Proline

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP18A041 Certificate TypeCertificate of Analysis Date07/02/2025 Catalog NumberA13137.22,A13137.14,A13137.06
Lot NumberQ11K025 Certificate TypeCertificate of Analysis Date06/05/2024 Catalog NumberA13137.22,A13137.14,A13137.06
Lot Number10184620 Certificate TypeCertificate of Analysis Date06/06/2023 Catalog NumberA13137.14
Lot NumberF22X001 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13137.22,A13137.14,A13137.06
Lot Number10179537 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13137.22,A13137.14,A13137.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DL-Proline, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.