missing translation for 'onlineSavingsMsg'
Learn More

DL-Prolinol, 98%, Thermo Scientific Chemicals

Product Code. 15408858
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

DL-Prolinol is used as asymmetric catalysts in proline organ catalysis reactions, and it is also used in many pharmaceutical, biotechnological applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Prolinol is used as asymmetric catalysts in proline organ catalysis reactions, and it is also used in many pharmaceutical, biotechnological applications.

Solubility
Soluble in ethanol.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 498-63-5
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD00601073
InChI Key HVVNJUAVDAZWCB-UHFFFAOYSA-N
Synonym 2-hydroxymethyl pyrrolidine, 2-pyrrolidinemethanol, prolinol, pyrrolidin-2-yl-methanol, dl-prolinol, 2-hydroxymethylpyrrolidine, pyrrolidin-2-yl methanol, 2-pyrrolidinylmethanol, pyrrolidine-2-methanol, 2-pyrrolidinylmethanol #
PubChem CID 90908
ChEBI CHEBI:59652
IUPAC Name pyrrolidin-2-ylmethanol
SMILES C1CC(NC1)CO

Specifications

Refractive Index 1.484
Boiling Point 76°C (2mmHg)
Quantity 5 g
Solubility Information Soluble in ethanol.
Formula Weight 101.15
Percent Purity 98%
Chemical Name or Material DL-Prolinol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'