missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Prolinol, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
DL-Prolinol is used as asymmetric catalysts in proline organ catalysis reactions, and it is also used in many pharmaceutical, biotechnological applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
DL-Prolinol is used as asymmetric catalysts in proline organ catalysis reactions, and it is also used in many pharmaceutical, biotechnological applications.
Solubility
Soluble in ethanol.
Notes
Keep container tightly closed. Store away from oxidizing agents.
DL-Prolinol is used as asymmetric catalysts in proline organ catalysis reactions, and it is also used in many pharmaceutical, biotechnological applications.
Solubility
Soluble in ethanol.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
Specifications
Refractive Index | 1.484 |
Boiling Point | 76°C (2mmHg) |
Quantity | 25 g |
Solubility Information | Soluble in ethanol. |
Formula Weight | 101.15 |
Percent Purity | 98% |
Chemical Name or Material | DL-Prolinol |
Safety and Handling
Product Identifier
- DL-Prolinol
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : DL-Prolinol
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'