missing translation for 'onlineSavingsMsg'
Learn More
DL-tert-Leucine, 95%, Thermo Scientific Chemicals Product Code.: 15439438

DL-tert-Leucine, 95%, Thermo Scientific Chemicals

Product Code. 15439438
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15439438

Brand: Thermo Scientific Alfa Aesar H63007.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33105-81-6
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00065933
InChI Key NPDBDJFLKKQMCM-UHFFFAOYSA-N
Synonym dl-tert-leucine, dl-tert-butylglycine, tert-butylglycine, h-dl-tle-oh, tert-leucine, 3-methylvaline, 2-amino-3,3-dimethyl-butyric acid, 3-methyl valine, t-butylglycine, dl-t-butylglycine
PubChem CID 306131
ChEBI CHEBI:72770
IUPAC Name 2-amino-3,3-dimethylbutanoic acid
SMILES CC(C)(C)C(N)C(O)=O

Specifications

Melting Point >300°C (decomposition)
Beilstein 1721823
Quantity 5 g
Formula Weight 131.18
Percent Purity 95%
Physical Form Solid
Chemical Name or Material DL-tert-Leucine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DL-tert-Leucine, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.