missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals DL-Threonine, 99% Product Code.: 11443294

Thermo Scientific Chemicals DL-Threonine, 99%

Product Code. 11443294
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11443294

Brand: Thermo Scientific Chemicals A10606.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

DL-Threonine is used as a polar essential amino acid. It serves as a precursor of glycine as well as used in the biosynthesis of proteins. Further, it is used for biochemical research. In addition to this, it is used as a nutrition enhancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Threonine is used as a polar essential amino acid. It serves as a precursor of glycine as well as used in the biosynthesis of proteins. Further, it is used for biochemical research. In addition to this, it is used as a nutrition enhancer.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-68-2
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00063722
InChI Key AYFVYJQAPQTCCC-PWNYCUMCSA-N
Synonym dl-threonine, allo-dl-threonine, threonine, dl, dl-allothreonine, dl-2-amino-3-hydroxybutanoic acid, threonine l, h-dl-thr-oh, dl-allo-threonine, allothreonine, d, wln: qy1&yzvq-lShow MoreShow Less
PubChem CID 205
ChEBI CHEBI:38263
SMILES C[C@@H](O)[C@@H](N)C(O)=O

Specifications

Melting Point ∼244°C (decomposition)
Beilstein 1721647
Merck Index 14,9380
Solubility Information Soluble in water.
IUPAC Name 2-amino-3-hydroxybutanoic acid
Quantity 25 g
Formula Weight 119.12
Percent Purity 99%
Chemical Name or Material DL-Threonine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464354 Certificate TypeCertificate of Analysis Date16/08/2024 Catalog NumberA10606.22,A10606.14,A10606.36
Lot NumberA0437899 Certificate TypeCertificate of Analysis Date19/04/2024 Catalog NumberA10606.22,A10606.14,A10606.36
Lot NumberA0437900 Certificate TypeCertificate of Analysis Date26/09/2023 Catalog NumberA10606.22,A10606.14,A10606.36
Lot Number10239846 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA10606.22,A10606.14,A10606.36
Lot Number10239845 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA10606.22,A10606.14,A10606.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals DL-Threonine, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.