missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals DL-Threonine, 99.5% Product Code.: 10347020

Thermo Scientific Chemicals DL-Threonine, 99.5%

Product Code. 10347020
25 g, Glass Bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10347020

Brand: Thermo Scientific Chemicals 138940250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-68-2
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00063722
InChI Key AYFVYJQAPQTCCC-PWNYCUMCSA-N
Synonym dl-threonine, allo-dl-threonine, threonine, dl, dl-allothreonine, dl-2-amino-3-hydroxybutanoic acid, threonine l, h-dl-thr-oh, dl-allo-threonine, allothreonine, d, wln: qy1&yzvq-l
PubChem CID 205
ChEBI CHEBI:38263
SMILES C[C@@H](O)[C@@H](N)C(O)=O

Specifications

Arsenic (As) 2ppm max.
Color White
Chlorine (Cl) 200ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Assay Percent Range 99.5%
Loss on Drying 0.2% max. (1 g, 105°C, 3 hrs)
Other Amino Acids (TLC) Passes Test
Packaging Glass Bottle
Linear Formula CH3CH(OH)CH(NH2)CO2H
Beilstein 04, 514
Merck Index 10, 9229
Sulfated Ash 0.1% max.
IUPAC Name (2R,3R)-2-amino-3-hydroxybutanoic acid
Quantity 25 g
Formula Weight 119.12
Percent Purity 98.0 to 101.0% (on dried substance)
Physical Form Crystalline Powder or Crystals
Chemical Name or Material DL-Threonine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466310 Certificate TypeCertificate of Analysis Date23/09/2024 Catalog Number138940250,138945000,138941000
Lot NumberA0437899 Certificate TypeCertificate of Analysis Date15/07/2024 Catalog Number138940250,138945000,138941000
Lot NumberA0464354 Certificate TypeCertificate of Analysis Date11/07/2024 Catalog Number138940250,138945000,138941000
Lot NumberA0463395 Certificate TypeCertificate of Analysis Date05/06/2024 Catalog Number138940250,138945000,138941000
Lot NumberA0400884 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number138940250,138945000,138941000
Product Identifier
  • DL-Threonine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals DL-Threonine, 99.5% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.