missing translation for 'onlineSavingsMsg'
Learn More

DL-Tryptophanamide, 98%, Thermo Scientific Chemicals

Product Code. 15499808
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6720-02-1
Molecular Formula C11H13N3O
Molecular Weight (g/mol) 203.25
MDL Number MFCD03826622
InChI Key JLSKPBDKNIXMBS-UHFFFAOYNA-N
PubChem CID 89366
IUPAC Name 2-amino-3-(1H-indol-3-yl)propanamide
SMILES NC(CC1=CNC2=CC=CC=C12)C(N)=O

Specifications

Color White
Synonym 2-amino-3-1h-indol-3-yl propanamide, h-dl-trp-nh2, alpha-amino-1h-indole-3-propanamide, tryptophan amide, dl-tryptophanamide, d,l-tryptophanamide, l-tryptophanamide, 9, h-dl-trp-nh hcl, dl-trytophan amide hydrochloride, 1h-indole-3-propanamide,a-amino
Formula Weight 203.24
Percent Purity 98%
Quantity 250 mg
Physical Form Crystalline Powder
Chemical Name or Material DL-Tryptophanamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'