missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals DL-Tyrosine, 98% Product Code.: 11494764

Thermo Scientific Chemicals DL-Tyrosine, 98%

Product Code. 11494764
10 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11494764

Brand: Thermo Scientific Chemicals A13740.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The tyrosine of this diet was labeled by adding to it an amount of synthetic, isotopic dl-tyrosine cor- responding to twice that already present in the food. Incorporation of total C14 into muscle protein was reduced in 186-day infected mice fed dl-tyrosine-2-C-14 as compared to muscle proteins obtained from noninfected animals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The tyrosine of this diet was labeled by adding to it an amount of synthetic, isotopic dl-tyrosine cor- responding to twice that already present in the food. Incorporation of total C14 into muscle protein was reduced in 186-day infected mice fed dl-tyrosine-2-C-14 as compared to muscle proteins obtained from noninfected animals.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 556-03-6
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD00063074
InChI Key OUYCCCASQSFEME-UHFFFAOYNA-N
Synonym dl-tyrosine, h-dl-tyr-oh, 2-amino-3-4-hydroxyphenyl propanoic acid, tyrosin, tyrosine, dl, l-tyrosine, free base, tirosina, l-tryosine, 3-4-hydroxyphenyl-dl-alanine, benzenepropanoic acid, s
PubChem CID 1153
ChEBI CHEBI:18186
IUPAC Name 2-amino-3-(4-hydroxyphenyl)propanoic acid
SMILES NC(CC1=CC=C(O)C=C1)C(O)=O

Specifications

Melting Point ∼325°C (decomposition)
Beilstein 515881
Merck Index 14,9839
Solubility Information Soluble in water.
Quantity 10 g
Formula Weight 181.19
Percent Purity 98%
Chemical Name or Material DL-Tyrosine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10247738 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog NumberA13740.18,A13740.09,A13740.30
Lot NumberN07J002 Certificate TypeCertificate of Analysis Date10/02/2023 Catalog NumberA13740.18,A13740.09,A13740.30
Lot NumberX10I028 Certificate TypeCertificate of Analysis Date14/10/2022 Catalog NumberA13740.18,A13740.09,A13740.30
Lot NumberT18I038 Certificate TypeCertificate of Analysis Date27/07/2022 Catalog NumberA13740.18,A13740.09,A13740.30
Lot NumberN07I032 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA13740.18,A13740.09,A13740.30
compliance-icons
Product Identifier
  • DL-Tyrosine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : DL-Tyrosine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals DL-Tyrosine, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.