missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals DL-Tyrosine, 99% Product Code.: 10245640

Thermo Scientific Chemicals DL-Tyrosine, 99%

Product Code. 10245640
100 g, Glass Bottle
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10245640

Brand: Thermo Scientific Chemicals 163571000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 556-03-6
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD00063074
InChI Key OUYCCCASQSFEME-UHFFFAOYNA-N
Synonym dl-tyrosine, h-dl-tyr-oh, 2-amino-3-4-hydroxyphenyl propanoic acid, tyrosin, tyrosine, dl, l-tyrosine, free base, tirosina, l-tryosine, 3-4-hydroxyphenyl-dl-alanine, benzenepropanoic acid, sShow MoreShow Less
PubChem CID 1153
ChEBI CHEBI:18186
IUPAC Name 2-amino-3-(4-hydroxyphenyl)propanoic acid
SMILES NC(CC1=CC=C(O)C=C1)C(O)=O

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula 4-(HO)C6H4CH2CH(NH2)CO2H
Beilstein 14, 621
Merck Index 14, 9839
Quantity 100 g
Formula Weight 181.19
Percent Purity 98.5 to 101.5%
Physical Form Powder
Chemical Name or Material DL-Tyrosine, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0460043 Certificate TypeCertificate of Analysis Date08/02/2024 Catalog Number163570250,163571000
Lot NumberA0458279 Certificate TypeCertificate of Analysis Date04/12/2023 Catalog Number163570250,163571000
Lot NumberA0454117 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number163570250,163571000
Lot NumberA0369776 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number163570250,163571000
compliance-icons
Product Identifier
  • DL-Tyrosine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals DL-Tyrosine, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.