missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals DL-Valine, +99% Product Code.: 10428150

Thermo Scientific Chemicals DL-Valine, +99%

Product Code. 10428150
50 g, Plastic Bottle
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic Bottle
This item is not returnable. View return policy

Product Code. 10428150

Brand: Thermo Scientific Chemicals 172100500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For biochemistry
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 516-06-3
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00004267
InChI Key KZSNJWFQEVHDMF-UHFFFAOYSA-N
Synonym dl-valine, h-dl-val-oh, valin, valine, dl, dl-2-amino-3-methylbutanoic acid, 2-amino-3-methylbutanoic acid, dl, valina, dl-.alpha.-aminoisovaleric acid, dl-2-amino-3-methylbutyric acid, 2-aminoisovaleric acid
PubChem CID 1182
ChEBI CHEBI:27266
IUPAC Name 2-amino-3-methylbutanoic acid
SMILES CC(C)C(N)C(O)=O

Specifications

Melting Point 283.5°C to 285.0°C
Color White
Chlorine (Cl) 400ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Loss on Drying 0.2% max. (105°C, 3 hrs)
Packaging Plastic Bottle
Linear Formula (CH3)2CHCH(NH2)CO2H
Beilstein 04, 430
Merck Index 14, 9909
Transmittance 95% min. (c=10, 2N HCl, 430nm) 1cm cell
Sulfated Ash 0.2% max.
Solubility Information Solubility in water: 68g/L. Other solubilities: practically insoluble in cold alcohol and ether, soluble in mineral acids, insoluble in organic solvents, slightly soluble in benzene, ethanol
Density 1.3100g/mL
Quantity 50 g
Formula Weight 117.15
Percent Purity ≥99%
Specific Gravity 1.31
Grade Biochemical
Physical Form Crystalline Powder
Chemical Name or Material DL-Valine, 99+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0427987 Certificate TypeCertificate of Analysis Date11/09/2023 Catalog Number172100010,172102500,172100500
Lot NumberA0454075 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number172100010,172102500,172100500
Lot NumberA0422360 Certificate TypeCertificate of Analysis Date04/09/2020 Catalog Number172102500,172100500
Product Identifier
  • DL-Valine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals DL-Valine, +99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.