missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals DL-Valine, 99% Product Code.: 11482768

Thermo Scientific Chemicals DL-Valine, 99%

Product Code. 11482768
2500 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11482768

Brand: Thermo Scientific Chemicals A16756.0E

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 516-06-3
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00004267
InChI Key KZSNJWFQEVHDMF-UHFFFAOYSA-N
Synonym dl-valine, h-dl-val-oh, valin, valine, dl, dl-2-amino-3-methylbutanoic acid, 2-amino-3-methylbutanoic acid, dl, valina, dl-.alpha.-aminoisovaleric acid, dl-2-amino-3-methylbutyric acid, 2-aminoisovaleric acid
PubChem CID 1182
ChEBI CHEBI:27266
IUPAC Name 2-amino-3-methylbutanoic acid
SMILES CC(C)C(N)C(O)=O

Specifications

Melting Point ∼296°C (sublimation)
Beilstein 506689
Merck Index 14,9909
Quantity 2500 g
Formula Weight 117.15
Percent Purity 99%
Chemical Name or Material DL-Valine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250916 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA16756.22,A16756.36,A16756.0E
Lot Number10251956 Certificate TypeCertificate of Analysis Date09/08/2024 Catalog NumberA16756.22,A16756.36,A16756.0E
Lot Number10248542 Certificate TypeCertificate of Analysis Date05/12/2023 Catalog NumberA16756.22,A16756.36,A16756.0E
Lot Number10244873 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberA16756.22,A16756.36,A16756.0E
Lot NumberT29I020 Certificate TypeCertificate of Analysis Date20/09/2022 Catalog NumberA16756.22,A16756.36,A16756.0E

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals DL-Valine, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.