Learn More
DLin-KC2-DMA, MedChemExpress
MedChemExpress DLin-KC2-DMA is a cationic/ionizable lipid for siRNA delivery.
Brand:  Medchem Express HY-112758
Encompass_Preferred
 View more versions of this product
Product Code. 16450987
  • 746.55 € / 50mg
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
DLin-KC2-DMA is a cationic/ionizable lipid for siRNA delivery.
Specifications
Specifications
DLin-KC2-DMA
Colorless
C43H79NO2
Ethanol : 100 mg/mL (155.74 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (155.74 mM; Need ultrasonic)
642.09
98.0%
Liquid
1190197-97-7
Research
50 mg
CCCCC/C=C\C/C=C\CCCCCCCCC1(CCCCCCCC/C=C\C/C=C\CCCCC)OC(CCN(C)C)CO1
642.09
Research