missing translation for 'onlineSavingsMsg'
Learn More

Leica Microsystems DM IL LED Microscope

Product Code. p-8498005
Click to view available options
For Use With (Application):
Brightfield and Fluorescence Microscopy
This item is not returnable. View return policy

Product Code. 12878035

Brand: Leica Microsystems DMILLEDLF

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ergonomically designed inverted microscope with high-performance optics. Leica Microsystems™ DM IL LED Microscope is ideal for cell culture examination and micromanipulation.

  • Ideal for applications in cell biology and medicine, micromanipulation, biotechnology, developmental biology, transgenics, molecular biology and fluorescence.
  • Ergonomic arrangement of controls for fatigue free operation.
  • In addition to phase contrast and brightfield illumination, the DM IL features Leica's unique integrated modulation contrast (IMC) without the need for special objectives.
    • Available for 10x, 20x, 32x, and 40x magnification.
  • Comprehensive set of contrast methods.
  • LED illumination provides low energy consumption and no heat build up.
  • LEDs last 50,000 hours.
  • Fluorescence capable (LF Model)
  • Sturdy design with low stage height and plenty of operation space.
  • Long working distances to accommodate large culture flasks.
  • Can be upgraded to accommodate large-scale cell production.
  • Certified for in-vitro diagnostics (IVD) including in-vitro fertilization (IVF).

Specifications

Includes Fluorescence slider with three positions for filter cubes, manual light stop, fluorescence LED Leica SFL100, 50 W Hg, 100 W Hg, Leica EL6000 fiber optic coupling.
Voltage 230 V
For Use With (Application) Brightfield and Fluorescence Microscopy
Electrical Requirements 230 V 50/60 Hz
Frequency 50/60 Hz
Product Type Inverted Microscope
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.