missing translation for 'onlineSavingsMsg'
Learn More

DMAP1 Mouse anti-Human, Unconjugated, Clone: 1A4E8, Proteintech

Mouse Monoclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen DMAP1
Clone 1A4E8
Concentration 1000 μg/mL
Applications Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16809685
View Documents
Proteintech
67183-1-IG-20UL
20 μL
145.00 €
20µL
Please to purchase this item. Need a web account? Register with us today!
16899675
View Documents
Proteintech
67183-1-IG-150UL
150 μL
410.00 €
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The DMAP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DMAP1. This antibody recognizes human, mouse, rat antigen. The DMAP1 antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Specifications

DMAP1
1000 μg/mL
Monoclonal
Mouse
55929
Primary
1A4E8
Western Blot, ELISA
Unconjugated
Human, Rat, Mouse
IgG1
Videos
SDS
Documents

Documents

Product Certifications