missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ DMEM/F-12, HEPES

DMEM/F-12, HEPES
29.22 € - 272.00 €
Specifications
Concentration | 1 X |
---|---|
Form | Liquid |
Product Type | DMEM/F-12 Medium |
With Additives | Glutamine, HEPES, Phenol Red, Sodium Pyruvate |
For Use With (Application) | Mammalian cell culture |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11594426
![]() |
Gibco™
31330038 |
500 mL |
29.22 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11504436
![]() |
Gibco™
31330095 |
10 x 500 mL |
272.00 €
5000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dulbecco's Modified Eagle Medium:Nutrient Mixture F-12 (DMEM / F-12) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM / F-12 include MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts. We offer a variety of Gibco™ DMEM / F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool. Gibco™ DMEM / F-12 is a 1:1 mixture of DMEM and Ham's F-12. This formulation combines DMEM's high concentrations of glucose, amino acids, and vitamins with F-12's wide variety of components.Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ DMEM / F-12 is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ DMEM / F-12 product made in our Grand Island facility (11330-032). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
DMEM / F-12 contains no proteins, lipids, or growth factors. Therefore, DMEM / F-12 may require supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM / F-12 uses a sodium bicarbonate buffer system and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Specifications
1 X | |
DMEM/F-12 Medium | |
Mammalian cell culture | |
6.9 to 7.3 | |
2 to 8°C |
Liquid | |
Glutamine, HEPES, Phenol Red, Sodium Pyruvate | |
ISO 13485 standards, FDA | |
For in-vitro diagnostic use | |
DMEM/F-12 Medium, 500mL, 1 X Concentration, Liquid Form, With L-glutamine, HEPES, Phenol Red, Sodium Pyruvate, 6.9 to 7.3 pH, 12 Months Shelf Life, 270 to 330mOsm/kg Osmolality, 2 to 8°C Storage Conditions, Used for Mammalian Cell Culture |
Safety and Handling
missing translation for 'shelfLife' : 12 Months
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ DMEM/F-12, HEPES