missing translation for 'onlineSavingsMsg'
Learn More

DMNPE-caged ATP diammonium salt, Tocris Bioscience™

Product Code. 30002046
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Caged ATP

DMNPE-caged ATP; quantum yield is 0.07. Brain penetrant.

Specifications

Molecular Formula C20H27N6O17P3·2NH3
Quantity 10 mg
Synonym Adenosine 5'-triphosphate P''-[1-(4, 5-dimethoxy-2-nitrophenyl)ethyl] ester diammonium salt
Formula Weight 750.44
Chemical Name or Material DMNPE-caged ATP diammonium salt
missing translation for 'provideContentCorrection'

missing translation for 'type03FormHeader'

missing translation for 'productTitle'

missing translation for 'thankYou' missing translation for 'feedbackSubmitted'