missing translation for 'onlineSavingsMsg'
Learn More

DMSO, Dimethyl Sulfoxide, GC Headspace Grade, Thermo Scientific™

Product Code. 15572393
1 L, Glass Bottle
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

DMSO processed to high purity for accurate and repeatable determination of trace levels of residual solvents.

  • Every lot is tested to ensure interference free baselines for interference free determination of Class 1, 2, and 3 residual solvents
  • 1 ppm max. of Class 1 Solvents, 10 ppm max. of Class 2 solvents, and 50 ppm max of Class 3 residual solvents
  • This product is UV Absorbance Tested to ensure low organic contamination
  • Processed for low water content to facilitate the extraction of organic volatile impurities, or VOC
TRUSTED_SUSTAINABILITY

Specifications

Absorbance 0.40 AU Maximum (270nm), 0.18 AU Maximum (290nm), 0.06 AU Maximum (310nm), 0.02 AU Maximum (330nm), 0.01 AU Maximum (350 to 400nm)
CAS 67-68-5
Color Colorless
Quantity 1 L
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.01 % Maximum
Molecular Formula C2H6OS
Titratable Acid 0.001 mEq/g Maximum
MDL Number MFCD00002089
Water 0.05% Maximum
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
SMILES CS(C)=O
IUPAC Name methylsulfinylmethane
Molecular Weight (g/mol) 78.13
PubChem CID 679
ChEBI CHEBI:28262
Formula Weight 78.13
Percent Purity ≥99.9%
Grade GC Headspace
Chemical Name or Material DMSO, Dimethyl Sulfoxide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DMSO, Dimethyl Sulfoxide, GC Headspace Grade, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.