missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ DNA LoBind™ Tubes
GREENER_CHOICE

Unleash the power of precision in your DNA research with Eppendorf DNA LoBind Tubes. Engineered with proprietary LoBind technology, these microcentrifuge tubes redefine sample handling, minimizing the risk of loss and maximizing DNA recovery. Ideal for low-concentration applications, including PCR, qPCR, and DNA sequencing, these tubes ensure accuracy, purity, and optimal yields.

Brand:  Eppendorf™ 0030108035

30.40 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10686313

  • 32.08 € / Pack of 250
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Surface-coating-free, including substances like silicone, plasticizers, biocides or mold release agents to reduce the risk of sample interference.
  • Lot-certified PCR clean purity grade guarantees freedom from human DNA, DNase, RNase, and PCR inhibitors.
  • Available in tube, microplate, and deep-well plate formats for convenient upscaling of experiments.
  • Precision lid sealing minimizes evaporation rates in the tube format.
  • Suitable for molecular biology applications with a centrifugation speed rating of up to 30,000 x g (25,000 x g for 2.0 mL tube).
  • Snap, screw, and SnapTec™ cap options available.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DNA, DNase, RNase, PCR Inhibitors free
Transparent, Clear
0.5 mL
Polypropylene
Safe-lock lid
250 Tubes (5 Bags x 50 Tubes)
Non-sterile
Snap Cap
Yes
Cylindrical
PCR Inhibitor Free
DNA LoBind consumables are perfect for DNA or RNA applications in forensic analysis, microarrays, next generation sequencing, gene cloning and general molecular biology
30,000 x g
HDPE, Polypropylene
0.5 mL
250 Tubes (5 Bags of 50)
DNA or RNA applications in forensic analysis, microarrays, next generation sequencing, gene cloning and general molecular biology
250 Pk.
PCR Clean, PCR Clean (Non-sterile)
30,000 x g
Yes
Autoclavable
0.02 oz.
Conical
Microcentrifuge Tube
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.