Learn More
Eppendorf™ DNA LoBind™ Tubes
Unleash the power of precision in your DNA research with Eppendorf DNA LoBind Tubes. Engineered with proprietary LoBind technology, these microcentrifuge tubes redefine sample handling, minimizing the risk of loss and maximizing DNA recovery. Ideal for low-concentration applications, including PCR, qPCR, and DNA sequencing, these tubes ensure accuracy, purity, and optimal yields.
17.72 € - 32.08 €
Specifications
Certifications/Compliance | DNA, DNase, RNase, PCR Inhibitors free |
---|---|
Closure Material | HDPE, Polypropylene |
Color | Transparent, Clear |
Material | Polypropylene |
For Use With | DNA or RNA applications in forensic analysis, microarrays, next generation sequencing, gene cloning and general molecular biology |
Product Code | Brand | Capacity (Metric) | Max. RCF | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Max. RCF | Price | Quantity & Availability | ||||
10051232
|
Eppendorf™
0030108051 |
1.5 mL | 30,000 x g |
17.72 €
Pack of 250 |
In Stock
Log in to see stock. |
||||
10686313
|
Eppendorf™
0030108035 |
0.5 mL | 30,000 x g |
32.08 €
Pack of 250 |
In Stock
Log in to see stock. |
||||
10031282
|
Eppendorf™
0030108078 |
2 mL | 25,000 x g |
21.23 €
Pack of 250 |
In Stock
Log in to see stock. |
||||
Description
- Surface-coating-free, including substances like silicone, plasticizers, biocides or mold release agents to reduce the risk of sample interference.
- Lot-certified PCR clean purity grade guarantees freedom from human DNA, DNase, RNase, and PCR inhibitors.
- Available in tube, microplate, and deep-well plate formats for convenient upscaling of experiments.
- Precision lid sealing minimizes evaporation rates in the tube format.
- Suitable for molecular biology applications with a centrifugation speed rating of up to 30,000 x g (25,000 x g for 2.0 mL tube).
- Snap, screw, and SnapTec™ cap options available.
Specifications
DNA, DNase, RNase, PCR Inhibitors free | |
Transparent, Clear | |
DNA or RNA applications in forensic analysis, microarrays, next generation sequencing, gene cloning and general molecular biology | |
250 Pk. | |
Non-sterile | |
Yes | |
Cylindrical | |
Conical | |
Microcentrifuge Tube |
HDPE, Polypropylene | |
Polypropylene | |
Safe-lock lid | |
250 Tubes (5 Bags x 50 Tubes) | |
Yes | |
Autoclavable | |
PCR Inhibitor Free | |
Snap Cap | |
PCR Clean, PCR Clean (Non-sterile) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.