Learn More
MP Biomedicals™ Q-Bio Taq DNA polymerase
MP Biomedicals™ Q-Bio Taq DNA polymerase is a thermostable DNA polymerase designed for mutliplex PCR and RAPDs.
Brand: MP Biomedicals™ 11EPQBT025
This item is not returnable.
View return policy
Description
- Q-Bio Taq DNA Polymerase is encoded by a modified form of the Thermus aquaticus DNA Polymerase gene with a N-terminal deletion.
- No 5′-3′ exonuclease activity
- Q-Bio Taq DNA Polymerase lacks the 5′exonuclease activity, which makes this enzyme particularly suitable for Multiplex PCR, by using several pairs of primers.
- Highly thermostable
- Q-Bio Taq DNA Polymerase is more thermostable than full-length Taq DNA Polymerase, as repeated exposure to 98°C does not diminish the enzyme activity.
- Cannot degrade the 5′ end of primers
- Q-Bio Taq DNA Polymerase has not the ability to degrade the 5′ end of primers. This is why this enzyme is particularly suitable for Multiplex PCR and RAPDs.
Specifications
Highly thermostable | |
Multiplex PCR and RAPD | |
98°C |