missing translation for 'onlineSavingsMsg'
Learn More

DNase I from Bovine Pancreas, Thermo Scientific™

Derived from Bovine pancreas.

101.76 €

Specifications

Product Type Cell Lysis Enzyme
Donor Source Bovine Pancreas
Form Lyophilized
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15845278
View Documents
Thermo Scientific
J14340.F+
20 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Synonym: E.C.3.1.21.1
  • MDL: MFCD00130917
Specifications

Specifications

Cell Lysis Enzyme
Bovine Pancreas
Lyophilized
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.