missing translation for 'onlineSavingsMsg'
Learn More

DNQX disodium salt, Tocris Bioscience™

Product Code. 30001300
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective non-NMDA iGluR antagonist; water-soluble salt of DNQX (Cat. No. 0189)

DNQX disodium salt is a more water-soluble form of DNQX (Cat. No. 0189). DNQX is a selective non-NMDA glutamate receptor antagonist. (IC50 = 0.1 μM and 0.5 μM for kainate and AMPA receptors, respectively). DNQX is a neuroleptic agent that shows pro-oxidative properties. DNQX selectively depolarizes rat thalamic reticular nucleus neurons.

Chemical Identifiers

CAS 1312992-24-7
Molecular Formula C8H2N4Na2O6
Molecular Weight (g/mol) 296.106
InChI Key GPSBSOYURFUVKJ-UHFFFAOYSA-L
Synonym dnqx disodium salt, dnqx disodium, 6,7-dinitroquinoxaline-2,3-dione disodium salt
PubChem CID 45073428
IUPAC Name disodium;6,7-dinitroquinoxaline-2,3-diolate
SMILES C1=C2C(=CC(=C1[N+](=O)[O-])[N+](=O)[O-])N=C(C(=N2)[O-])[O-].[Na+].[Na+]

Specifications

Quantity 10 mg
Formula Weight 296.1
Percent Purity >98%
Chemical Name or Material DNQX disodium salt
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'