Learn More
Tocris Bioscience™ Docosahexaenoic acid
RXR agonist
Brand:  Tocris Bioscience™ 3687/100
146.97 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18095412
  • 185.00 € / 100mg
Explore more special offers
Description
Description
Docosahexaenoic acid is a endogenous omega-3 fatty acid. Acts as a selective retinoid X receptor (RXR) agonist that displays no activity at RAR, thyroid hormone receptor or the vitamin D receptor (VDR). Activates all three RXR isoforms. Also shown to inhibit Aβ1-42 fibrillation and toxicity in vitro.
Specifications
Specifications
>98%
Docosahexaenoic acid
Docosahexaenoic acid
100 mg
328.49