Learn More
Dodecanedioyl dichloride, 98%, Thermo Scientific Chemicals
40.10 € - 151.00 €
Chemical Identifiers
CAS | 4834-98-4 |
---|---|
Molecular Formula | C12H20Cl2O2 |
Molecular Weight (g/mol) | 267.2 |
MDL Number | MFCD00012243 |
InChI Key | CNXXEPWXNDFGIG-UHFFFAOYSA-N |
Synonym | dodecanedioyldichloride, dodecandioyldichloride, dodecanedioyl chloride, acmc-209kct, dodecanedioic acid dichloride, 1,12-dodecanedioyl dichloride, dodecanedioyl dichloride |
PubChem CID | 2733242 |
IUPAC Name | dodecanedioyl dichloride |
SMILES | C(CCCCCC(=O)Cl)CCCCC(=O)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10028701
|
Thermo Scientific Acros
305340050 |
5 g | Glass bottle |
40.10 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10686671
|
Thermo Scientific Acros
305340250 |
25 g | Glass bottle |
151.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4834-98-4 | |
267.2 | |
CNXXEPWXNDFGIG-UHFFFAOYSA-N | |
2733242 | |
C(CCCCCC(=O)Cl)CCCCC(=O)Cl |
C12H20Cl2O2 | |
MFCD00012243 | |
dodecanedioyldichloride, dodecandioyldichloride, dodecanedioyl chloride, acmc-209kct, dodecanedioic acid dichloride, 1,12-dodecanedioyl dichloride, dodecanedioyl dichloride | |
dodecanedioyl dichloride |
Specifications
4834-98-4 | |
1.0600g/mL | |
>110°C | |
97.5% min. (GC) | |
ClCO(CH2)10COCl | |
MFCD00012243 | |
dodecanedioyldichloride, dodecandioyldichloride, dodecanedioyl chloride, acmc-209kct, dodecanedioic acid dichloride, 1,12-dodecanedioyl dichloride, dodecanedioyl dichloride | |
C(CCCCCC(=O)Cl)CCCCC(=O)Cl | |
267.2 | |
267.2 | |
Liquid |
Yellow to Brown | |
140°C (0.5 mmHg) | |
Authentic | |
C12H20Cl2O2 | |
1.4670 to 1.469 | |
1.06 | |
CNXXEPWXNDFGIG-UHFFFAOYSA-N | |
dodecanedioyl dichloride | |
2733242 | |
98% | |
Dodecanedioyl dichloride, 98% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
Recommended Storage : Product may darken during storage
RUO – Research Use Only