Learn More
Thermo Scientific Chemicals Dodecyl sulfate sodium salt, 99+%, for molecular biology, DNAse, RNAse and Protease free
126.63 € - 1324.59 €
Specifications
Absorbance | (0.1 M in water), (0.1 M in water) at 0nm, 0.05 max. at 260nm, 0.05 max. at 280nm |
---|---|
Chemical Name or Material | Dodecyl sulfate, sodium salt |
Color | White |
CAS | 151-21-3, 7757-82-6, 7647-14-5 |
Physical Form | Powder |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10733124
|
Thermo Scientific Chemicals
327311000 |
100 g |
126.63 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10524441
|
Thermo Scientific Chemicals
327315000 |
500 g |
397.98 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11984561
|
Thermo Scientific Chemicals
327310025 |
2.5 kg |
1324.59 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
(0.1 M in water), (0.1 M in water) at 0nm, 0.05 max. at 260nm, 0.05 max. at 280nm | |
White | |
Powder | |
>150°C | |
99+% | |
CH3(CH2)11OSO3Na | |
Dodecyl Sodium Sulfate, SDS, Sodium lauryl sulfate | |
DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
288.38 | |
CHEBI:8984 | |
≥99% |
Dodecyl sulfate, sodium salt | |
151-21-3, 7757-82-6, 7647-14-5 | |
206.0°C | |
Authentic | |
C12H25NaO4S | |
MFCD00036175 | |
Solubility in water: 130g/L (20°C). Other solubilities: partly soluble in alcohol, soluble 75g/L in ethanol (20°C) | |
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O | |
3423265 | |
288.38 | |
Molecular Biology |
Safety and Handling
GHS H Statement
Flammable solid. May cause respiratory irritation. Harmful if swallowed. Harmful if inhaled. Causes serious eye damage. Causes skin irritation. Harmful to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Danger
missing translation for 'einecsNumber' : 205-788-1
missing translation for 'tsca' : TSCA
- Dodecyl sulfate sodium salt
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.