missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Dog BSEP Vesicles

Dog BSEP Vesicles are insect derived purified plasma membranes with inserted BSEP transport protein (bile salt export pump, ABCB11).

Brand:  Gibco™ GM0019

Product Code. 10501051

  • 2105.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Our Dog BSEP Vesicles can be used to evaluate test compounds and drug interactions with BSEP in in vitro assays. BSEP is an ATP-binding cassette (ABC) transporter that plays a key role in bile acid excretion from the liver.

Use Dog BSEP Vesicles to:

• Investigate the transporter interactions of your drug candidates
• Assess potential for transporter-mediated drug-drug interactions
• Obtain high quality results with a large signal to noise ratios

Concept Behind ABC Transporter Vesicles
ABC transporter vesicles are easy-to-use, efficient reagents for early assessment of a drug candidate's substrate and drug interaction potential. While ABC transporters typically mediate the export of substrates out of cells, transporters expressed on these inside-out vesicles import substrates into the vesicles. It is therefore possible to quantitatively evaluate transport activity for your compound by determining the amount incorporated into the vesicles.

Clear and Reliable Results
Prepared from Sf9 cells which have been engineered to over-express specific ABC transporters, these 'inside-out' vesicles provide high levels of transporter activity with low background, giving you a clear signal if your compound is a substrate or inhibitor of a specific efflux transporter.

Transporter Vesicle Details
Dog BSEP Vesicles are part of a larger group of transporter vesicles known as ABC transporter vesicles. They are prepared from purified plasma membranes isolated from an insect cell system (Sf9 cells transfected with baculovirus) expressing BSEP. Evaluation of the inhibitory activity of newly developed compounds towards BSEP-mediated transport of bile acids (e.g. taurocholate) may give an indication of potential hepatotoxicity of the compounds.

For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Frozen
Research Grade
Dog Vesicle
Vial
0.5 mL
Canine
Store at -80°C.
Dry Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberXA8JX35 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog NumberGM0019
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Dog BSEP Vesicles > Store at -80°C.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.