missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dolichos Biflorus (Horse Gram) Agglutinin (DBA), fluorescein (FITC)

Bright-green DBA-fluorophor conjugate

Brand:  Invitrogen™ L32474

110.88 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.


Product Code. 17116642

  • 168.00 € / 2mg
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Dolichos Biflorus (Horse Gram) Agglutinin (DBA), fluorescein (FITC), is a bright-green DBA-fluorophor conjugate. DBA has been used to establish secretor status in blood-group-A individuals by hemagglutination inhibition techniques and for blood typing.
  • This lectin has also been used as a general marker of developing renal collecting ducts. It has also been used as a marker of cell surface change in murine embryonic stem cells during early differentiation.
  • Very bright FITC conjugate detected with traditional FITC/GFP filter set, Ex/Em: 495/515 nm
  • Applications: immunofluorescence, glycobiology
  • Sugar specificity: N-acetylgalactosamine
  • Inhibiting/eluting sugar: 200 mM N-acetylgalactosamine
  • Blood group specificity: A1 >>A2
  • Unconjugated lectin molecular weight: 111 KDa
  • Immunofluorescence working concentration: 5–20 μg/mL
  • DBA-FITC is provided at a 2 mg/mL concentration in 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.08% sodium azide, 0.1 mM Ca2+. If a precipitate forms upon long-term storage at 4°C, warm to 37°C and centrifuge before use.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FITC
Horse gram
Approved for shipment at Room Temperature or on Wet Ice
Lectins
2 mg
When stored at or below 2–8°C in the dark, the product is stable for 6 months after reciept.

Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.