missing translation for 'onlineSavingsMsg'
Learn More

Domnick Hunter Pure air desiccant dryer DAS4 220V 50Hz 227L/min Domnick Hunter

Brand:  Domnick Hunter DAS4

Pure air desiccant dryer DAS4 220V 50Hz 227L/min Domnick Hunter

Code : 04

Additional Details : Weight : 24.00000kg

Product Code. 12822726

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers