Learn More
Donkey anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight™ 550, Invitrogen™
Donkey Polyclonal Secondary Antibody
Brand:  Invitrogen SA510027
Product Code. 13487177
  • 183.00 € / 500µg
Explore more special offers
Description
Description
This antibody is cross-adsorbed and exhibits minimum reactivity to bovine, chicken, goat, human, mouse, rabbit and sheep.
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rat IgG (H+L) Cross-Adsorbed
Polyclonal
DyLight 550
Donkey
Antigen affinity chromatography
RUO
Rat
Liquid
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
0.5 mg/mL
PBS with 0.2% BSA and 0.09% sodium azide; pH 6.8 to 7.4
Rat IgG-heavy and light chain
500 μg
Secondary
4° C