Learn More
Dopamine hydrochloride, 99%, Thermo Scientific Chemicals
31.86 € - 438.18 €
Chemical Identifiers
CAS | 62-31-7 |
---|---|
Molecular Formula | C8H12ClNO2 |
Molecular Weight (g/mol) | 189.64 |
MDL Number | MFCD00012898 |
InChI Key | CTENFNNZBMHDDG-UHFFFAOYSA-N |
Synonym | dopamine hydrochloride, 3-hydroxytyramine hydrochloride, dopamine hcl, 4-2-aminoethyl benzene-1,2-diol hydrochloride, intropin, dopastat, revivan, dynatra, 3,4-dihydroxyphenethylamine hydrochloride, cardiosteril |
PubChem CID | 65340 |
IUPAC Name | 4-(2-aminoethyl)benzene-1,2-diol;hydrochloride |
SMILES | [H+].[Cl-].NCCC1=CC=C(O)C(O)=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11499033
|
Thermo Scientific Alfa Aesar
A11136.06 |
5 g |
31.86 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11409043
|
Thermo Scientific Alfa Aesar
A11136.14 |
25 g |
117.59 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15403037
|
Thermo Scientific Alfa Aesar
A11136.22 |
100 g |
438.18 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A neurotransmitter and vasopressor that modulates cortical activation. Dopamine hydrochloride is a vasopressor that moderates cortical activation. It is used as a precursor to norepinepherine and epinephrine. It is an important neurotransmitter and chemical messenger that helps in the transmission of signals in the brain and other vital areas.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA neurotransmitter and vasopressor that modulates cortical activation. Dopamine hydrochloride is a vasopressor that moderates cortical activation. It is used as a precursor to norepinepherine and epinephrine. It is an important neurotransmitter and chemical messenger that helps in the transmission of signals in the brain and other vital areas.
Solubility
Soluble in water, methanol and dimethyl sulfoxide. Insoluble in ether, petroleum ether, chloroform, benzene and toluene.
Notes
Store in cool place. Incompatible with strong oxidizing agents.
Chemical Identifiers
62-31-7 | |
189.64 | |
CTENFNNZBMHDDG-UHFFFAOYSA-N | |
65340 | |
[H+].[Cl-].NCCC1=CC=C(O)C(O)=C1 |
C8H12ClNO2 | |
MFCD00012898 | |
dopamine hydrochloride, 3-hydroxytyramine hydrochloride, dopamine hcl, 4-2-aminoethyl benzene-1,2-diol hydrochloride, intropin, dopastat, revivan, dynatra, 3,4-dihydroxyphenethylamine hydrochloride, cardiosteril | |
4-(2-aminoethyl)benzene-1,2-diol;hydrochloride |
Specifications
62-31-7 | |
99% | |
MFCD00012898 | |
Hygroscopic; Light sensitive | |
dopamine hydrochloride, 3-hydroxytyramine hydrochloride, dopamine hcl, 4-2-aminoethyl benzene-1,2-diol hydrochloride, intropin, dopastat, revivan, dynatra, 3,4-dihydroxyphenethylamine hydrochloride, cardiosteril | |
CTENFNNZBMHDDG-UHFFFAOYSA-N | |
4-(2-aminoethyl)benzene-1,2-diol;hydrochloride | |
65340 | |
99% |
243°C to 250°C | |
C8H12ClNO2 | |
3656720 | |
14,3421 | |
Soluble in water,methanol and dimethyl sulfoxide. Insoluble in ether,petroleum ether,chloroform,benzene and toluene. | |
[H+].[Cl-].NCCC1=CC=C(O)C(O)=C1 | |
189.64 | |
189.64 | |
Dopamine hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P312-P321-P405-P403+P233-P501a
Warning
missing translation for 'einecsNumber' : 200-527-8
missing translation for 'rtecsNumber' : UX1092000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only